Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAATACAGATCGTTTTAATGTGC[C/T]GTAAATATTTCTCGCACAGCATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605977 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IMMP2L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IMMP2L - inner mitochondrial membrane peptidase subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244606.1 | 778 | UTR 3 | NP_001231535.1 | |||
NM_032549.3 | 778 | UTR 3 | NP_115938.1 | |||
XM_005250630.3 | 778 | UTR 3 | XP_005250687.1 | |||
XM_011516605.2 | 778 | UTR 3 | XP_011514907.1 | |||
XM_011516608.2 | 778 | Intron | XP_011514910.1 | |||
XM_011516609.2 | 778 | UTR 3 | XP_011514911.1 | |||
XM_011516611.2 | 778 | Intron | XP_011514913.1 | |||
XM_011516613.2 | 778 | UTR 3 | XP_011514915.1 | |||
XM_017012699.1 | 778 | Intron | XP_016868188.1 | |||
XM_017012700.1 | 778 | Intron | XP_016868189.1 | |||
XM_017012701.1 | 778 | Intron | XP_016868190.1 | |||
XM_017012702.1 | 778 | Intron | XP_016868191.1 | |||
XM_017012703.1 | 778 | Intron | XP_016868192.1 | |||
XM_017012704.1 | 778 | Intron | XP_016868193.1 | |||
XM_017012705.1 | 778 | UTR 3 | XP_016868194.1 |