Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATCTGATGTATTTTTGTCATTT[C/T]GCCTCAAGCGGTAATTTTTATAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182392 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCN7A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCN7A - sodium voltage-gated channel alpha subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002976.3 | 5352 | Missense Mutation | CAA,CGA | Q,R 1643 | NP_002967.2 | |
XM_006712680.2 | 5352 | Missense Mutation | CAA,CGA | Q,R 1643 | XP_006712743.1 | |
XM_006712681.3 | 5352 | Missense Mutation | CAA,CGA | Q,R 1643 | XP_006712744.1 | |
XM_006712682.3 | 5352 | Missense Mutation | CAA,CGA | Q,R 1643 | XP_006712745.1 | |
XM_011511615.2 | 5352 | Missense Mutation | CAA,CGA | Q,R 1643 | XP_011509917.1 | |
XM_017004667.1 | 5352 | Missense Mutation | CAA,CGA | Q,R 1643 | XP_016860156.1 |