Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCACGCTCCTACGCGGCGGCTTG[G/T]GTTTCGCAGGCGGTTGGGGATCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608016 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COA4 - cytochrome c oxidase assembly factor 4 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016565.2 | 94 | UTR 5 | NP_057649.2 | |||
XM_017017883.1 | 94 | Intron | XP_016873372.1 | |||
XM_017017884.1 | 94 | UTR 5 | XP_016873373.1 |
PAAF1 - proteasomal ATPase associated factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267803.1 | 94 | Intron | NP_001254732.1 | |||
NM_001267804.1 | 94 | Intron | NP_001254733.1 | |||
NM_001267805.1 | 94 | Intron | NP_001254734.1 | |||
NM_001267806.1 | 94 | Intron | NP_001254735.1 | |||
NM_025155.2 | 94 | Intron | NP_079431.1 | |||
XM_005274308.2 | 94 | Intron | XP_005274365.2 | |||
XM_005274309.1 | 94 | Intron | XP_005274366.1 | |||
XM_017018357.1 | 94 | Intron | XP_016873846.1 | |||
XM_017018358.1 | 94 | Intron | XP_016873847.1 | |||
XM_017018359.1 | 94 | Intron | XP_016873848.1 |