Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGAGTATAAGAATTCACTGGATA[A/G]CTACTGGAACTTTTTTGCATGTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
H2AFV PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
H2AFV - H2A histone family member V | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012412.4 | Intron | NP_036544.1 | ||||
NM_138635.3 | Intron | NP_619541.1 | ||||
NM_201436.2 | Intron | NP_958844.1 | ||||
NM_201516.2 | Intron | NP_958924.1 | ||||
NM_201517.2 | Intron | NP_958925.1 |