Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGATCTCAGCTACCCAGGCATC[A/G]GCCAGGCAGTACTGCAGGAGCTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607732 MIM: 611672 | ||||||||||||||||||||
Literature Links: |
SARM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SARM1 - sterile alpha and TIR motif containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC46A1 - solute carrier family 46 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242366.2 | 1088 | Silent Mutation | GCC,GCT | A,A 330 | NP_001229295.1 | |
NM_080669.5 | 1088 | Silent Mutation | GCC,GCT | A,A 330 | NP_542400.2 | |
XM_005277786.3 | 1088 | Silent Mutation | GCC,GCT | A,A 330 | XP_005277843.1 | |
XM_017024110.1 | 1088 | Silent Mutation | GCC,GCT | A,A 256 | XP_016879599.1 |