Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGCCACAGCTCACCTGGCGGCAC[C/T]GCAGCGTCTGGCCCAGCTGGCCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607465 MIM: 614642 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDAN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDAN1 - codanin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138477.2 | 3251 | Missense Mutation | CAG,CGG | Q,R 1065 | NP_612486.2 | |
XM_005254176.4 | 3251 | Missense Mutation | CAG,CGG | Q,R 1066 | XP_005254233.1 | |
XM_011521270.2 | 3251 | Missense Mutation | CAG,CGG | Q,R 1074 | XP_011519572.1 | |
XM_011521271.2 | 3251 | Missense Mutation | CAG,CGG | Q,R 1073 | XP_011519573.1 | |
XM_011521274.2 | 3251 | Missense Mutation | CAG,CGG | Q,R 729 | XP_011519576.1 |
STARD9 - StAR related lipid transfer domain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |