Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGTGAAGATGGCTGCCTCTGCC[G/T]TTGCTGGTGCAGTGAGAGCAGCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600378 MIM: 611841 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IMMT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IMMT - inner membrane mitochondrial protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4779 - microRNA 4779 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL35 - mitochondrial ribosomal protein L35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016622.3 | 74 | Missense Mutation | GTT,TTT | V,F 6 | NP_057706.2 | |
NM_145644.2 | 74 | Missense Mutation | GTT,TTT | V,F 6 | NP_663619.1 | |
XM_005264361.2 | 74 | Intron | XP_005264418.1 |