Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTGCTACTTCCTGGAGACTCAG[A/G]ACTGCTCCTTCCACTAGAATCAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605196 MIM: 616653 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COQ3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COQ3 - coenzyme Q3, methyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PNISR - PNN interacting serine and arginine rich protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322405.1 | 2446 | Missense Mutation | CCT,TCT | P,S 762 | NP_001309334.1 | |
NM_001322406.1 | 2446 | Missense Mutation | CCT,TCT | P,S 762 | NP_001309335.1 | |
NM_001322408.1 | 2446 | Missense Mutation | CCT,TCT | P,S 762 | NP_001309337.1 | |
NM_001322410.1 | 2446 | Missense Mutation | CCT,TCT | P,S 762 | NP_001309339.1 | |
NM_001322412.1 | 2446 | Missense Mutation | CCT,TCT | P,S 762 | NP_001309341.1 | |
NM_001322413.1 | 2446 | Missense Mutation | CCT,TCT | P,S 762 | NP_001309342.1 | |
NM_001322414.1 | 2446 | Missense Mutation | CCT,TCT | P,S 744 | NP_001309343.1 | |
NM_001322415.1 | 2446 | Missense Mutation | CCT,TCT | P,S 744 | NP_001309344.1 | |
NM_001322416.1 | 2446 | Intron | NP_001309345.1 | |||
NM_001322417.1 | 2446 | Intron | NP_001309346.1 | |||
NM_001322418.1 | 2446 | Intron | NP_001309347.1 | |||
NM_001322419.1 | 2446 | Intron | NP_001309348.1 | |||
NM_015491.2 | 2446 | Missense Mutation | CCT,TCT | P,S 762 | NP_056306.1 | |
NM_032870.3 | 2446 | Missense Mutation | CCT,TCT | P,S 762 | NP_116259.2 | |
XM_005266912.3 | 2446 | Missense Mutation | CCT,TCT | P,S 762 | XP_005266969.2 | |
XM_017010710.1 | 2446 | Missense Mutation | CCT,TCT | P,S 744 | XP_016866199.1 | |
XM_017010711.1 | 2446 | Missense Mutation | CCT,TCT | P,S 744 | XP_016866200.1 | |
XM_017010712.1 | 2446 | Missense Mutation | CCT,TCT | P,S 667 | XP_016866201.1 | |
XM_017010713.1 | 2446 | Missense Mutation | CCT,TCT | P,S 667 | XP_016866202.1 |