Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTGAACACTGGAGGAGGAAGAC[A/T]GTTATCCAGCAGACAAGAGCTAAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610545 MIM: 610546 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGFL2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IGFL2 - IGF like family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002915.2 | 2497 | Intron | NP_001002915.2 | |||
NM_001135113.1 | 2497 | Intron | NP_001128585.1 | |||
XM_011526501.1 | 2497 | Intron | XP_011524803.1 | |||
XM_017026335.1 | 2497 | Intron | XP_016881824.1 |
IGFL3 - IGF like family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207393.1 | 2497 | Silent Mutation | ACA,ACT | T,T 15 | NP_997276.1 | |
XM_011526953.2 | 2497 | Silent Mutation | ACA,ACT | T,T 15 | XP_011525255.1 | |
XM_017026796.1 | 2497 | Silent Mutation | ACA,ACT | T,T 15 | XP_016882285.1 | |
XM_017026797.1 | 2497 | Silent Mutation | ACA,ACT | T,T 15 | XP_016882286.1 |