Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTCGTTCTGTTAATTTAGGCTT[A/G]ACACTTTAACCATTTCAAGCACCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606498 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MS4A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MS4A3 - membrane spanning 4-domains A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031666.1 | Intron | NP_001026836.1 | ||||
NM_001031809.1 | Intron | NP_001026979.1 | ||||
NM_006138.4 | Intron | NP_006129.4 | ||||
XM_011545363.2 | Intron | XP_011543665.1 |