Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCCAAATTAGAGGCCATTTCAA[C/T]ATGGAATTCTGGGGTGACAATTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609546 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP29 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
USP29 - ubiquitin specific peptidase 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZIM3 - zinc finger imprinted 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052882.1 | 1873 | UTR 3 | NP_443114.1 |