Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCGGGCTGCTCCGTGCATCTTTC[A/C]CCCCAGGCGTCAGGAACTGCGCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DZANK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR3192 - microRNA 3192 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR3F - RNA polymerase III subunit F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282526.1 | 91 | UTR 5 | NP_001269455.1 | |||
NM_006466.3 | 91 | UTR 5 | NP_006457.2 | |||
XM_011529141.1 | 91 | UTR 5 | XP_011527443.1 | |||
XM_017027596.1 | 91 | UTR 5 | XP_016883085.1 |