Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCGAGGCCCAGCAGGAGAGGAG[A/C]CTGGGTCTCCACGCTCACAAAGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALCOCO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CALCOCO1 - calcium binding and coiled-coil domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143682.1 | 1911 | Missense Mutation | GGC,GTC | G,V 476 | NP_001137154.1 | |
NM_020898.2 | 1911 | Missense Mutation | GGC,GTC | G,V 561 | NP_065949.1 | |
XM_011538601.1 | 1911 | Missense Mutation | GGC,GTC | G,V 561 | XP_011536903.1 | |
XM_011538602.1 | 1911 | Missense Mutation | GGC,GTC | G,V 561 | XP_011536904.1 | |
XM_011538603.1 | 1911 | Missense Mutation | GGC,GTC | G,V 561 | XP_011536905.1 | |
XM_011538604.1 | 1911 | Missense Mutation | GGC,GTC | G,V 561 | XP_011536906.1 | |
XM_017019707.1 | 1911 | Missense Mutation | GGC,GTC | G,V 561 | XP_016875196.1 | |
XM_017019708.1 | 1911 | Missense Mutation | GGC,GTC | G,V 238 | XP_016875197.1 | |
XM_017019709.1 | 1911 | Missense Mutation | GGC,GTC | G,V 238 | XP_016875198.1 |