Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTAAGCTGAAGGTGTGACTACA[A/G]ATTATTGATGAACTTCCCTCAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613481 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC62 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC62 - coiled-coil domain containing 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_201435.4 | Intron | NP_958843.2 | ||||
XM_006719643.1 | Intron | XP_006719706.1 | ||||
XM_006719644.1 | Intron | XP_006719707.1 | ||||
XM_011538856.1 | Intron | XP_011537158.1 | ||||
XM_011538857.1 | Intron | XP_011537159.1 | ||||
XM_011538859.1 | Intron | XP_011537161.1 | ||||
XM_017020033.1 | Intron | XP_016875522.1 |
Set Membership: |
HapMap |