Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615788 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
N4BP2L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
N4BP2L2 - NEDD4 binding protein 2 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278432.1 | 3056 | Missense Mutation | AAT,AGT | N,S 529 | NP_001265361.1 | |
NM_001320836.1 | 3056 | Missense Mutation | AAT,AGT | N,S 973 | NP_001307765.1 | |
NM_014887.2 | 3056 | Intron | NP_055702.1 | |||
NM_033111.4 | 3056 | Missense Mutation | AAT,AGT | N,S 544 | NP_149102.3 | |
XM_011534869.2 | 3056 | Missense Mutation | AAT,AGT | N,S 529 | XP_011533171.1 | |
XM_011534870.2 | 3056 | Missense Mutation | AAT,AGT | N,S 457 | XP_011533172.1 | |
XM_011534871.2 | 3056 | Missense Mutation | AAT,AGT | N,S 457 | XP_011533173.1 | |
XM_011534872.2 | 3056 | Missense Mutation | AAT,AGT | N,S 427 | XP_011533174.1 | |
XM_011534874.2 | 3056 | Intron | XP_011533176.1 | |||
XM_017020324.1 | 3056 | Missense Mutation | AAT,AGT | N,S 973 | XP_016875813.1 | |
XM_017020325.1 | 3056 | Missense Mutation | AAT,AGT | N,S 973 | XP_016875814.1 | |
XM_017020326.1 | 3056 | Missense Mutation | AAT,AGT | N,S 973 | XP_016875815.1 | |
XM_017020327.1 | 3056 | Missense Mutation | AAT,AGT | N,S 973 | XP_016875816.1 | |
XM_017020328.1 | 3056 | Missense Mutation | AAT,AGT | N,S 973 | XP_016875817.1 | |
XM_017020329.1 | 3056 | Missense Mutation | AAT,AGT | N,S 973 | XP_016875818.1 | |
XM_017020330.1 | 3056 | Missense Mutation | AAT,AGT | N,S 973 | XP_016875819.1 | |
XM_017020331.1 | 3056 | Missense Mutation | AAT,AGT | N,S 560 | XP_016875820.1 | |
XM_017020332.1 | 3056 | Missense Mutation | AAT,AGT | N,S 560 | XP_016875821.1 | |
XM_017020333.1 | 3056 | Missense Mutation | AAT,AGT | N,S 560 | XP_016875822.1 | |
XM_017020334.1 | 3056 | Missense Mutation | AAT,AGT | N,S 560 | XP_016875823.1 | |
XM_017020335.1 | 3056 | Missense Mutation | AAT,AGT | N,S 529 | XP_016875824.1 | |
XM_017020336.1 | 3056 | Missense Mutation | AAT,AGT | N,S 457 | XP_016875825.1 | |
XM_017020337.1 | 3056 | Missense Mutation | AAT,AGT | N,S 457 | XP_016875826.1 | |
XM_017020338.1 | 3056 | Missense Mutation | AAT,AGT | N,S 457 | XP_016875827.1 | |
XM_017020339.1 | 3056 | Missense Mutation | AAT,AGT | N,S 427 | XP_016875828.1 | |
XM_017020340.1 | 3056 | Missense Mutation | AAT,AGT | N,S 427 | XP_016875829.1 | |
XM_017020341.1 | 3056 | Missense Mutation | AAT,AGT | N,S 427 | XP_016875830.1 | |
XM_017020342.1 | 3056 | Intron | XP_016875831.1 | |||
XM_017020343.1 | 3056 | Missense Mutation | AAT,AGT | N,S 375 | XP_016875832.1 | |
XM_017020344.1 | 3056 | Missense Mutation | AAT,AGT | N,S 375 | XP_016875833.1 | |
XM_017020345.1 | 3056 | Missense Mutation | AAT,AGT | N,S 375 | XP_016875834.1 | |
XM_017020346.1 | 3056 | Missense Mutation | AAT,AGT | N,S 375 | XP_016875835.1 | |
XM_017020347.1 | 3056 | Intron | XP_016875836.1 | |||
XM_017020348.1 | 3056 | Intron | XP_016875837.1 | |||
XM_017020349.1 | 3056 | Intron | XP_016875838.1 |