Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCGGAAACCACAGAGCTGCTGCT[G/T]CTTGGAGCAGGGCCCCCACGGAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609595 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RSPO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RSPO1 - R-spondin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001038633.3 | 1376 | Missense Mutation | AAG,CAG | K,Q 162 | NP_001033722.1 | |
NM_001242908.1 | 1376 | Missense Mutation | AAG,CAG | K,Q 162 | NP_001229837.1 | |
NM_001242909.1 | 1376 | Missense Mutation | AAG,CAG | K,Q 135 | NP_001229838.1 | |
NM_001242910.1 | 1376 | Intron | NP_001229839.1 | |||
XM_006710583.3 | 1376 | Missense Mutation | AAG,CAG | K,Q 162 | XP_006710646.1 |