Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTATGTGTAGTCATTGTTCCAC[C/T]AGTATCTCACTTAATGTTCAGCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610462 MIM: 603733 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RTN4RL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RTN4RL2 - reticulon 4 receptor-like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC43A1 - solute carrier family 43 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198810.1 | Intron | NP_001185739.1 | ||||
NM_003627.5 | Intron | NP_003618.1 | ||||
XM_005274358.4 | Intron | XP_005274415.1 | ||||
XM_011545320.2 | Intron | XP_011543622.1 | ||||
XM_011545321.2 | Intron | XP_011543623.1 | ||||
XM_011545322.1 | Intron | XP_011543624.1 | ||||
XM_017018451.1 | Intron | XP_016873940.1 | ||||
XM_017018452.1 | Intron | XP_016873941.1 | ||||
XM_017018453.1 | Intron | XP_016873942.1 |