Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGGTGGGTTGGGCTGGGAGCAG[A/G]GGTCCTACTGGGGTGCTGAGTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607264 MIM: 613939 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNA1G-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACNA1G-AS1 - CACNA1G antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EPN3 - epsin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105371824 - uncharacterized LOC105371824 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA20 - spermatogenesis associated 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258372.1 | Intron | NP_001245301.1 | ||||
NM_001258373.1 | Intron | NP_001245302.1 | ||||
NM_022827.3 | Intron | NP_073738.2 |