Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 184756 MIM: 615519 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SREBF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SREBF1 - sterol regulatory element binding transcription factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOM1L2 - target of myb1 like 2 membrane trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033551.2 | 5070 | UTR 3 | NP_001028723.1 | |||
NM_001082968.1 | 5070 | UTR 3 | NP_001076437.1 | |||
NM_001288786.1 | 5070 | UTR 3 | NP_001275715.1 | |||
NM_001288787.1 | 5070 | UTR 3 | NP_001275716.1 | |||
NM_001288788.1 | 5070 | UTR 3 | NP_001275717.1 | |||
NM_001288789.1 | 5070 | UTR 3 | NP_001275718.1 | |||
XM_005256461.2 | 5070 | UTR 3 | XP_005256518.1 | |||
XM_005256462.1 | 5070 | UTR 3 | XP_005256519.1 | |||
XM_005256463.2 | 5070 | UTR 3 | XP_005256520.1 | |||
XM_005256466.1 | 5070 | UTR 3 | XP_005256523.1 | |||
XM_011523662.1 | 5070 | UTR 3 | XP_011521964.1 | |||
XM_017024201.1 | 5070 | UTR 3 | XP_016879690.1 |