Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGCAGAGGAGCCTGTCGTGGCC[A/G]GCGTCCTCCAGTGCCCCGGGAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608382 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJA3 - DnaJ heat shock protein family (Hsp40) member A3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NMRAL1 - NmrA-like family domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305141.1 | Intron | NP_001292070.1 | ||||
NM_001305142.1 | Intron | NP_001292071.1 | ||||
NM_020677.4 | Intron | NP_065728.1 | ||||
XM_006720905.3 | Intron | XP_006720968.1 | ||||
XM_006720906.2 | Intron | XP_006720969.1 | ||||
XM_011522567.2 | Intron | XP_011520869.1 | ||||
XM_017023485.1 | Intron | XP_016878974.1 | ||||
XM_017023486.1 | Intron | XP_016878975.1 | ||||
XM_017023487.1 | Intron | XP_016878976.1 | ||||
XM_017023488.1 | Intron | XP_016878977.1 | ||||
XM_017023489.1 | Intron | XP_016878978.1 | ||||
XM_017023490.1 | Intron | XP_016878979.1 | ||||
XM_017023491.1 | Intron | XP_016878980.1 | ||||
XM_017023492.1 | Intron | XP_016878981.1 |