Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAGAAAGGCAAGGATGTTTTTCT[A/G]TTTCTGGGGAATAAAAATGCTCTCG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615618 | |||||||||||||||||||||||
Literature Links: |
POGLUT1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
POGLUT1 - protein O-glucosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152305.2 | Intron | NP_689518.1 | ||||
XM_006713705.3 | Intron | XP_006713768.1 | ||||
XM_017006878.1 | Intron | XP_016862367.1 | ||||
XM_017006879.1 | Intron | XP_016862368.1 | ||||
XM_017006880.1 | Intron | XP_016862369.1 |
TMEM39A - transmembrane protein 39A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |