Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCGGGAGCTGCTGGAGGATTAT[C/T]GCTTTGGGCGGCAGCAGCTCGTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612419 MIM: 609435 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CILP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CILP2 - cartilage intermediate layer protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFA13 - NADH:ubiquinone oxidoreductase subunit A13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YJEFN3 - YjeF N-terminal domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190328.1 | 376 | Missense Mutation | CGC,TGC | R,C 36 | NP_001177257.1 | |
NM_198537.3 | 376 | Missense Mutation | CGC,TGC | R,C 86 | NP_940939.2 | |
XM_005259903.4 | 376 | Missense Mutation | CGC,TGC | R,C 159 | XP_005259960.1 | |
XM_011527996.2 | 376 | Missense Mutation | CGC,TGC | R,C 159 | XP_011526298.1 | |
XM_011527997.1 | 376 | UTR 5 | XP_011526299.1 | |||
XM_011527998.2 | 376 | UTR 5 | XP_011526300.1 | |||
XM_011527999.1 | 376 | UTR 5 | XP_011526301.1 | |||
XM_017026770.1 | 376 | Missense Mutation | CGC,TGC | R,C 39 | XP_016882259.1 | |
XM_017026771.1 | 376 | UTR 5 | XP_016882260.1 |