Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAACTTTGAGGCGAGGAAGATCC[C/T]GAGGGAAAGCCACACAGGTGAGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABARAPL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GABARAPL3 - GABA type A receptor associated protein like 3 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF774 - zinc finger protein 774 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004309.2 | 380 | Missense Mutation | CCG,CTG | P,L 65 | NP_001004309.2 | |
XM_017022127.1 | 380 | UTR 5 | XP_016877616.1 | |||
XM_017022128.1 | 380 | UTR 5 | XP_016877617.1 | |||
XM_017022129.1 | 380 | UTR 5 | XP_016877618.1 |
Set Membership: |
HapMap |