Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGGTAAACCAGTAATGAGCCAA[A/G]CATACGTACTCATGCAAATGTACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613133 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSPAN2 - tetraspanin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308315.1 | 803 | UTR 3 | NP_001295244.1 | |||
NM_001308316.1 | 803 | UTR 3 | NP_001295245.1 | |||
NM_005725.5 | 803 | UTR 3 | NP_005716.2 | |||
XM_016999996.1 | 803 | UTR 3 | XP_016855485.1 |