Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300364 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMCX3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARMCX3 - armadillo repeat containing, X-linked 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016607.3 | 1550 | Silent Mutation | AAA,AAG | K,K 296 | NP_057691.1 | |
NM_177947.2 | 1550 | Silent Mutation | AAA,AAG | K,K 296 | NP_808816.1 | |
NM_177948.2 | 1550 | Silent Mutation | AAA,AAG | K,K 296 | NP_808817.1 | |
XM_005262141.2 | 1550 | Silent Mutation | AAA,AAG | K,K 296 | XP_005262198.1 |
ARMCX6 - armadillo repeat containing, X-linked 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |