Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613383 MIM: 603692 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD54 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANKRD54 - ankyrin repeat domain 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138797.2 | 1602 | UTR 3 | NP_620152.1 | |||
XM_006724136.1 | 1602 | UTR 3 | XP_006724199.1 | |||
XM_006724137.1 | 1602 | UTR 3 | XP_006724200.1 | |||
XM_011529877.2 | 1602 | UTR 3 | XP_011528179.1 | |||
XM_011529878.2 | 1602 | UTR 3 | XP_011528180.1 | |||
XM_011529879.2 | 1602 | UTR 3 | XP_011528181.1 | |||
XM_011529880.2 | 1602 | UTR 3 | XP_011528182.1 | |||
XM_011529882.2 | 1602 | UTR 3 | XP_011528184.1 | |||
XM_011529883.2 | 1602 | UTR 3 | XP_011528185.1 | |||
XM_011529884.2 | 1602 | UTR 3 | XP_011528186.1 | |||
XM_017028590.1 | 1602 | UTR 3 | XP_016884079.1 | |||
XM_017028591.1 | 1602 | UTR 3 | XP_016884080.1 | |||
XM_017028593.1 | 1602 | UTR 3 | XP_016884082.1 |
GALR3 - galanin receptor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |