Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607320 MIM: 609293 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM98C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM98C - family with sequence similarity 98 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174905.3 | 209 | Missense Mutation | ACG,GCG | T,A 64 | NP_777565.3 | |
XM_017026354.1 | 209 | Missense Mutation | ACG,GCG | T,A 64 | XP_016881843.1 |
RASGRP4 - RAS guanyl releasing protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPRED3 - sprouty related EVH1 domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |