Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAACAGAAAGAAACACTGAAAAC[A/G]TATTTTTCGTTCATTGTCTTGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM41 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBM41 - RNA binding motif protein 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171080.1 | Intron | NP_001164551.1 | ||||
NM_001324242.1 | Intron | NP_001311171.1 | ||||
NM_001324243.1 | Intron | NP_001311172.1 | ||||
NM_001324244.1 | Intron | NP_001311173.1 | ||||
NM_018301.4 | Intron | NP_060771.3 | ||||
XM_011530980.2 | Intron | XP_011529282.1 | ||||
XM_011530981.2 | Intron | XP_011529283.1 |