Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCAGAGGGCACTTCCCCATCCA[A/G]CCCCACTCCCGTGTGCGATCCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606150 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FADS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FADS3 - fatty acid desaturase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB3IL1 - RAB3A interacting protein like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271686.1 | Intron | NP_001258615.1 | ||||
NM_013401.3 | Intron | NP_037533.2 | ||||
XM_005274140.4 | Intron | XP_005274197.1 | ||||
XM_006718647.3 | Intron | XP_006718710.1 | ||||
XM_011545196.2 | Intron | XP_011543498.1 | ||||
XM_011545197.2 | Intron | XP_011543499.1 | ||||
XM_011545198.2 | Intron | XP_011543500.1 | ||||
XM_011545199.2 | Intron | XP_011543501.1 | ||||
XM_011545200.2 | Intron | XP_011543502.1 | ||||
XM_011545201.2 | Intron | XP_011543503.1 |