Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGGGCCAGCAGATCCGGGAGGC[C/T]GGCGTGGAGGCGGCGGCGGATTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142957 MIM: 142950 MIM: 142956 MIM: 609688 | ||||||||||||||||||||
Literature Links: |
HOXA-AS3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HOXA-AS3 - HOXA cluster antisense RNA 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA10 - homeobox A10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA10-AS - HOXA10 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA10-HOXA9 - HOXA10-HOXA9 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA7 - homeobox A7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXA9 - homeobox A9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152739.3 | Intron | NP_689952.1 |
MIR196B - microRNA 196b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |