Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCGGCCCAGGATGGTCCACTTCC[C/G]GGCTGAGCGGGCCTTTCACCCAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604480 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RINL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RINL - Ras and Rab interactor like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195833.1 | 737 | Missense Mutation | CCG,CGG | P,R 219 | NP_001182762.1 | |
NM_198445.3 | 737 | Intron | NP_940847.1 | |||
XM_006723015.2 | 737 | Missense Mutation | CCG,CGG | P,R 219 | XP_006723078.1 | |
XM_006723016.2 | 737 | Missense Mutation | CCG,CGG | P,R 105 | XP_006723079.1 | |
XM_011526454.2 | 737 | Missense Mutation | CCG,CGG | P,R 219 | XP_011524756.1 | |
XM_011526455.2 | 737 | Missense Mutation | CCG,CGG | P,R 219 | XP_011524757.1 |
SIRT2 - sirtuin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |