Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGTACTGTAATTTAATAGAAGA[G/T]ATGGTCGTCTTAATATTTAATGATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603136 | ||||||||||||||||||||
Literature Links: |
CUL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CUL3 - cullin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257197.1 | 2786 | UTR 3 | NP_001244126.1 | |||
NM_001257198.1 | 2786 | UTR 3 | NP_001244127.1 | |||
NM_003590.4 | 2786 | UTR 3 | NP_003581.1 |