Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605097 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC45A3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC45A3 - solute carrier family 45 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033102.2 | 2101 | Silent Mutation | CTC,CTG | L,L 532 | NP_149093.1 | |
XM_005245556.3 | 2101 | Silent Mutation | CTC,CTG | L,L 532 | XP_005245613.1 | |
XM_005245557.4 | 2101 | Silent Mutation | CTC,CTG | L,L 532 | XP_005245614.1 | |
XM_005245560.2 | 2101 | Intron | XP_005245617.1 | |||
XM_017002638.1 | 2101 | Silent Mutation | CTC,CTG | L,L 532 | XP_016858127.1 |
Set Membership: |
HapMap |