Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCTTTGTGCTCCACCTATTGAA[C/T]TGGGCCCCACTGCTGCTCACCAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612472 MIM: 614032 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
METTL3 - methyltransferase like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019852.4 | Intron | NP_062826.2 | ||||
XM_006720206.3 | Intron | XP_006720269.1 | ||||
XM_011536968.1 | Intron | XP_011535270.1 |
TOX4 - TOX high mobility group box family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303523.1 | Intron | NP_001290452.1 | ||||
NM_014828.3 | Intron | NP_055643.1 |