Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACGTGCTGGGGGCCTGCTCACA[C/T]AGCTGCCGTACCACCTTCACCAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612035 MIM: 186975 | ||||||||||||||||||||
Literature Links: |
AARS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AARS2 - alanyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020745.3 | 2294 | Silent Mutation | CTA,CTG | L,L 903 | NP_065796.1 | |
XM_005249245.3 | 2294 | Silent Mutation | CTA,CTG | L,L 806 | XP_005249302.1 | |
XM_011514764.2 | 2294 | Silent Mutation | CTA,CTG | L,L 903 | XP_011513066.1 | |
XM_017011112.1 | 2294 | Silent Mutation | CTA,CTG | L,L 473 | XP_016866601.1 |
TCTE1 - t-complex-associated-testis-expressed 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |