Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609088 MIM: 604728 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL22 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FBXL22 - F-box and leucine rich repeat protein 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203373.2 | 599 | Intron | NP_976307.2 | |||
XM_005254320.3 | 599 | UTR 5 | XP_005254377.1 | |||
XM_006720478.1 | 599 | Intron | XP_006720541.1 | |||
XM_011521468.2 | 599 | Intron | XP_011519770.1 | |||
XM_011521469.1 | 599 | Intron | XP_011519771.1 | |||
XM_011521472.2 | 599 | Intron | XP_011519774.1 | |||
XM_017022086.1 | 599 | UTR 5 | XP_016877575.1 |
USP3 - ubiquitin specific peptidase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP3-AS1 - USP3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |