Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601194 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TLR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TLR1 - toll like receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003263.3 | 5974 | Intron | NP_003254.2 | |||
XM_005262662.4 | 5974 | UTR 3 | XP_005262719.1 | |||
XM_011513742.2 | 5974 | UTR 3 | XP_011512044.1 | |||
XM_011513743.2 | 5974 | UTR 3 | XP_011512045.1 | |||
XM_011513744.2 | 5974 | UTR 3 | XP_011512046.1 | |||
XM_011513745.2 | 5974 | UTR 3 | XP_011512047.1 | |||
XM_017008571.1 | 5974 | UTR 3 | XP_016864060.1 | |||
XM_017008572.1 | 5974 | UTR 3 | XP_016864061.1 |
Set Membership: |
HapMap |