Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615367 MIM: 614244 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NTAN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NTAN1 - N-terminal asparagine amidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270766.1 | 802 | Intron | NP_001257695.1 | |||
NM_001270767.1 | 802 | Intron | NP_001257696.1 | |||
NM_173474.3 | 802 | Intron | NP_775745.1 | |||
XM_011522355.2 | 802 | Missense Mutation | CCT,TCT | P,S 204 | XP_011520657.1 |
PDXDC1 - pyridoxal dependent decarboxylase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285444.1 | 802 | UTR 3 | NP_001272373.1 | |||
NM_001285445.1 | 802 | UTR 3 | NP_001272374.1 | |||
NM_001285447.1 | 802 | UTR 3 | NP_001272376.1 | |||
NM_001285448.1 | 802 | UTR 3 | NP_001272377.1 | |||
NM_001285449.1 | 802 | Intron | NP_001272378.1 | |||
NM_001285450.1 | 802 | Intron | NP_001272379.1 | |||
NM_001324019.1 | 802 | UTR 3 | NP_001310948.1 | |||
NM_001324020.1 | 802 | Intron | NP_001310949.1 | |||
NM_001324021.1 | 802 | Intron | NP_001310950.1 | |||
NM_015027.3 | 802 | UTR 3 | NP_055842.2 | |||
XM_005255173.1 | 802 | Intron | XP_005255230.1 | |||
XM_005255176.2 | 802 | Intron | XP_005255233.1 | |||
XM_006720865.2 | 802 | Intron | XP_006720928.2 | |||
XM_017023059.1 | 802 | Intron | XP_016878548.1 | |||
XM_017023060.1 | 802 | Intron | XP_016878549.1 | |||
XM_017023061.1 | 802 | Intron | XP_016878550.1 | |||
XM_017023062.1 | 802 | Intron | XP_016878551.1 | |||
XM_017023063.1 | 802 | Intron | XP_016878552.1 | |||
XM_017023064.1 | 802 | Intron | XP_016878553.1 | |||
XM_017023065.1 | 802 | Intron | XP_016878554.1 |
Set Membership: |
HapMap |