Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102560 MIM: 607643 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTG1 - actin gamma 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FSCN2 - fascin actin-bundling protein 2, retinal | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077182.2 | 907 | Intron | NP_001070650.1 | |||
NM_012418.3 | 907 | Intron | NP_036550.1 | |||
XM_011524587.2 | 907 | Intron | XP_011522889.1 | |||
XM_011524590.2 | 907 | UTR 5 | XP_011522892.1 |