Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGACCTTGTCTCAAAAAATAATA[A/C]GCCCCTGATCCCTAGCCTCTCCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103190 MIM: 611088 MIM: 610571 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARF3 - ADP ribosylation factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001659.2 | 2568 | Intron | NP_001650.1 | |||
XM_005268856.1 | 2568 | Intron | XP_005268913.1 | |||
XM_006719391.3 | 2568 | UTR 3 | XP_006719454.1 |
CCDC65 - coiled-coil domain containing 65 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FKBP11 - FK506 binding protein 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |