Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTATGACTAACAAGGGACTGAAA[A/G]GAAGCCATCCTACTGTCCTCATGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603317 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRQ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTPRQ - protein tyrosine phosphatase, receptor type Q | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145026.1 | 401 | Intron | NP_001138498.1 | |||
XM_017019273.1 | 401 | Missense Mutation | AGA,GGA | R,G 89 | XP_016874762.1 | |
XM_017019274.1 | 401 | Missense Mutation | AGA,GGA | R,G 89 | XP_016874763.1 | |
XM_017019275.1 | 401 | Missense Mutation | AGA,GGA | R,G 89 | XP_016874764.1 |
Set Membership: |
HapMap |