Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCTCCCCACGCTGGAGGCCGTGG[A/G]CCGCTCCCTTTAAAAGTTGAAACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609294 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABPB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABPB2 - GA binding protein transcription factor beta subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA6C - semaphorin 6C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178061.1 | 4115 | UTR 3 | NP_001171532.1 | |||
NM_001178062.1 | 4115 | UTR 3 | NP_001171533.1 | |||
NM_030913.4 | 4115 | UTR 3 | NP_112175.2 | |||
XM_005244835.3 | 4115 | UTR 3 | XP_005244892.1 | |||
XM_017000075.1 | 4115 | UTR 3 | XP_016855564.1 | |||
XM_017000076.1 | 4115 | UTR 3 | XP_016855565.1 | |||
XM_017000077.1 | 4115 | UTR 3 | XP_016855566.1 | |||
XM_017000078.1 | 4115 | UTR 3 | XP_016855567.1 | |||
XM_017000079.1 | 4115 | UTR 3 | XP_016855568.1 | |||
XM_017000080.1 | 4115 | UTR 3 | XP_016855569.1 | |||
XM_017000081.1 | 4115 | UTR 3 | XP_016855570.1 | |||
XM_017000082.1 | 4115 | UTR 3 | XP_016855571.1 |