Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTCTGTGTCTTTTAATTGGAGG[A/C]TTGAATCCATTTATATCCATGTTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PQLC2L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PQLC2L - PQ loop repeat containing 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099777.3 | Intron | NP_001093247.1 | ||||
NM_001130001.2 | Intron | NP_001123473.1 | ||||
NM_001130002.2 | Intron | NP_001123474.1 | ||||
NM_001243729.1 | Intron | NP_001230658.1 | ||||
XM_017005754.1 | Intron | XP_016861243.1 |