Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTATATTTGACATTTTTCATAGG[C/T]GTATTGTAAGAAAGGAAAGAAGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
55 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609602 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLF17 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLF17 - Kruppel like factor 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173484.3 | Intron | NP_775755.3 | ||||
XM_005270465.3 | Intron | XP_005270522.2 | ||||
XM_011540700.1 | Intron | XP_011539002.1 | ||||
XM_011540704.2 | Intron | XP_011539006.1 |
Set Membership: |
HapMap |