Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607249 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CATSPER2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CATSPER2 - cation channel sperm associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282309.1 | 384 | Missense Mutation | NP_001269238.1 | |||
NM_001282310.1 | 384 | Missense Mutation | ATA,GTA | I,V 63 | NP_001269239.1 | |
NM_054020.3 | 384 | Missense Mutation | ATA,GTA | I,V 57 | NP_473361.1 | |
NM_172095.2 | 384 | Missense Mutation | ATA,GTA | I,V 57 | NP_742093.1 | |
XM_017021904.1 | 384 | Missense Mutation | ATA,GTA | I,V 57 | XP_016877393.1 |
Set Membership: |
HapMap |