Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103390 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHNAK PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AHNAK - AHNAK nucleoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001620.2 | Intron | NP_001611.1 | ||||
NM_024060.3 | Intron | NP_076965.2 | ||||
XM_005274240.1 | Intron | XP_005274297.1 | ||||
XM_005274241.2 | Intron | XP_005274298.1 | ||||
XM_005274245.1 | Intron | XP_005274302.1 | ||||
XM_017018270.1 | Intron | XP_016873759.1 | ||||
XM_017018271.1 | Intron | XP_016873760.1 | ||||
XM_017018272.1 | Intron | XP_016873761.1 | ||||
XM_017018273.1 | Intron | XP_016873762.1 |