Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604061 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPT9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEPT9 - septin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113491.1 | Intron | NP_001106963.1 | ||||
NM_001113492.1 | Intron | NP_001106964.1 | ||||
NM_001113493.1 | Intron | NP_001106965.1 | ||||
NM_001113494.1 | Intron | NP_001106966.1 | ||||
NM_001113495.1 | Intron | NP_001106967.1 | ||||
NM_001113496.1 | Intron | NP_001106968.1 | ||||
NM_001293695.1 | Intron | NP_001280624.1 | ||||
NM_001293696.1 | Intron | NP_001280625.1 | ||||
NM_001293697.1 | Intron | NP_001280626.1 | ||||
NM_001293698.1 | Intron | NP_001280627.1 | ||||
NM_006640.4 | Intron | NP_006631.2 | ||||
XM_005256962.1 | Intron | XP_005257019.1 | ||||
XM_006721643.2 | Intron | XP_006721706.1 | ||||
XM_006721644.1 | Intron | XP_006721707.1 | ||||
XM_011524204.1 | Intron | XP_011522506.1 | ||||
XM_011524206.1 | Intron | XP_011522508.1 | ||||
XM_011524207.1 | Intron | XP_011522509.1 | ||||
XM_011524208.2 | Intron | XP_011522510.1 | ||||
XM_017024031.1 | Intron | XP_016879520.1 | ||||
XM_017024032.1 | Intron | XP_016879521.1 |