Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTAAACCTCTAATCCATAGATTG[C/T]AAATACAACGATAGCTTATTTTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602038 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DUSP8 - dual specificity phosphatase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004420.2 | 4702 | UTR 3 | NP_004411.2 | |||
XM_011519932.2 | 4702 | UTR 3 | XP_011518234.1 | |||
XM_011519933.2 | 4702 | UTR 3 | XP_011518235.1 |
LOC101927562 - uncharacterized LOC101927562 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005253274.3 | 4702 | Intron | XP_005253331.1 |
Set Membership: |
HapMap |