Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAATTGTATTTTCTTAGGCTATTG[A/T]TAAATGAATTATAGCTAAGCAGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608686 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYRFL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MYRFL - myelin regulatory factor-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182530.2 | 191 | Intron | NP_872336.2 | |||
XM_011539052.2 | 191 | UTR 5 | XP_011537354.1 | |||
XM_011539055.1 | 191 | Intron | XP_011537357.1 | |||
XM_017018967.1 | 191 | Intron | XP_016874456.1 | |||
XM_017018968.1 | 191 | UTR 5 | XP_016874457.1 | |||
XM_017018969.1 | 191 | Intron | XP_016874458.1 | |||
XM_017018970.1 | 191 | Intron | XP_016874459.1 | |||
XM_017018971.1 | 191 | UTR 5 | XP_016874460.1 | |||
XM_017018972.1 | 191 | UTR 5 | XP_016874461.1 | |||
XM_017018973.1 | 191 | Intron | XP_016874462.1 | |||
XM_017018974.1 | 191 | UTR 5 | XP_016874463.1 | |||
XM_017018975.1 | 191 | UTR 5 | XP_016874464.1 | |||
XM_017018976.1 | 191 | UTR 5 | XP_016874465.1 |
RAB3IP - RAB3A interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |