Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610142 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf29 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C12orf29 - chromosome 12 open reading frame 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009894.2 | Intron | NP_001009894.2 |
CEP290 - centrosomal protein 290 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025114.3 | Intron | NP_079390.3 | ||||
XM_011538756.2 | Intron | XP_011537058.1 | ||||
XM_011538757.2 | Intron | XP_011537059.1 | ||||
XM_011538758.2 | Intron | XP_011537060.1 | ||||
XM_011538759.2 | Intron | XP_011537061.1 | ||||
XM_011538760.1 | Intron | XP_011537062.1 | ||||
XM_011538761.1 | Intron | XP_011537063.1 | ||||
XM_011538762.2 | Intron | XP_011537064.1 | ||||
XM_011538763.2 | Intron | XP_011537065.1 | ||||
XM_011538764.2 | Intron | XP_011537066.1 | ||||
XM_011538765.2 | Intron | XP_011537067.1 | ||||
XM_011538766.2 | Intron | XP_011537068.1 | ||||
XM_017019980.1 | Intron | XP_016875469.1 | ||||
XM_017019981.1 | Intron | XP_016875470.1 | ||||
XM_017019982.1 | Intron | XP_016875471.1 | ||||
XM_017019983.1 | Intron | XP_016875472.1 |